Learn More
Cayman Chemical 2'-Fluoro-2'-deoxyadenosine, Purity: ≥98%, CAS: 64183-27-3, 500 mg
SDP
Catalog No. 502942090
Encompass_Preferred
A prodrug form of 2-F-adenine; reduces mean tumor weight in an E. coli purine nucleoside phosphorylase-expressing D54 glioblastoma mouse xenograft model at 100 mg/kg
Catalog No. 50-294-2090 Supplier Cayman Chemical Supplier No. 39923500 MG
A prodrug form of 2-F-adenine; reduces mean tumor weight in an E. coli purine nucleoside phosphorylase-expressing D54 glioblastoma mouse xenograft model at 100 mg/kg