Learn More
Cayman Chemical 2'-Fluoro-2'-deoxyadenosine, Purity: ≥98%, CAS: 64183-27-3, 5 g
SDP
Catalog No. 502941989
Encompass_Preferred
A prodrug form of 2-F-adenine; reduces mean tumor weight in an E. coli purine nucleoside phosphorylase-expressing D54 glioblastoma mouse xenograft model at 100 mg/kg
Catalog No. 50-294-1989 Supplier Cayman Chemical Supplier No. 399235 G
A prodrug form of 2-F-adenine; reduces mean tumor weight in an E. coli purine nucleoside phosphorylase-expressing D54 glioblastoma mouse xenograft model at 100 mg/kg