Learn More
Cayman Chemical 2'-Fluoro-2'-deoxyuridine, Purity: ≥98%, CAS: 784-71-4, 10 mg
SDP
Catalog No. 502942373
Encompass_Preferred
A derivative of uridine; hepatitis C polyU/UC RNA strands containing 2'-FdU bind to RIG-I but do not activate RIG-I signaling in a reporter assay using Huh7 cells; has been used as a starting material in the synthesis of RSV polymerases inhibitors
Catalog No. 50-294-2373 Supplier Cayman Chemical Supplier No. 4030210 MG
A derivative of uridine; hepatitis C polyU/UC RNA strands containing 2'-FdU bind to RIG-I but do not activate RIG-I signaling in a reporter assay using Huh7 cells; has been used as a starting material in the synthesis of RSV polymerases inhibitors