Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Fluoro-2'-deoxyuridine, 97%

Catalog No. AC455000010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455000010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455000010 Supplier Thermo Scientific Chemicals Supplier No. 455000010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 784-71-4
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD01317293
InChI Key UIYWFOZZIZEEKJ-XVFCMESISA-N
Synonym 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203
PubChem CID 150851
IUPAC Name 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Melting Point 151.0°C
Loss on Drying 1% max. (1 g, 105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 246.19
Percent Purity 97%
Chemical Name or Material 2'-Fluoro-2'-deoxyuridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of causing genetic defects. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.