Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one | 2301976-85-0 | 100.0% | 240.08 | C12H18BFO3 | 100 MG
SDP

Catalog No. 5000299732
Encompass_Preferred

2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one is a fluorinated cyclohexenone bearing a pinacol boronate (dioxaborolane) group. It is used as a versatile synthetic intermediate in medicinal chemistry and organic synthesis, enabling installation of aryl or vinyl groups through cross-coupling reactions.

  • Contains a pinacol boronate enabling Suzuki-Miyaura cross-coupling.
  • Fluorinated scaffold useful for modulating physicochemical and metabolic properties.
  • Synthetic intermediate suitable for late-stage functionalization in drug discovery.
  • Molecular formula C12H18BFO3 and molecular weight 240.08 g/mol.
  • Provided as a solid for research-scale synthesis; purity supplied on certificate of analysis.

Catalog No. 50-002-99732 Supplier Medchemexpress LLC Supplier No. HYW038588100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one is a fluorinated cyclohexenone bearing a pinacol boronate (dioxaborolane) group. It is used as a versatile synthetic intermediate in medicinal chemistry and organic synthesis, enabling installation of aryl or vinyl groups through cross-coupling reactions.

  • Contains a pinacol boronate enabling Suzuki-Miyaura cross-coupling.
  • Fluorinated scaffold useful for modulating physicochemical and metabolic properties.
  • Synthetic intermediate suitable for late-stage functionalization in drug discovery.
  • Molecular formula C12H18BFO3 and molecular weight 240.08 g/mol.
  • Provided as a solid for research-scale synthesis; purity supplied on certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.