Learn More
Medchemexpress LLC 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one | 2301976-85-0 | 98.0% | 240.08 g/mol | C12H18BFO3 | 250 MG
SDP
Catalog No. 5000303459
Encompass_Preferred
2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one is a boronic-ester functionalized cyclohexenone used as a synthetic intermediate in medicinal chemistry. It is intended for research and synthesis of active compounds, and is supplied with product documentation for laboratory use.
  • Boronic-ester functionalized cyclohexenone.
  • Used as a synthetic intermediate for medicinal chemistry.
  • Compatible with cross-coupling reactions such as Suzuki-Miyaura.
  • Offered in multiple research-scale pack sizes.
  • Supplied with product information and a datasheet for laboratory use.
Catalog No. 50-003-03459 Supplier Medchemexpress LLC Supplier No. 250MGHYW038588
Add to Cart
Add to Cart
2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one is a boronic-ester functionalized cyclohexenone used as a synthetic intermediate in medicinal chemistry. It is intended for research and synthesis of active compounds, and is supplied with product documentation for laboratory use.
  • Boronic-ester functionalized cyclohexenone.
  • Used as a synthetic intermediate for medicinal chemistry.
  • Compatible with cross-coupling reactions such as Suzuki-Miyaura.
  • Offered in multiple research-scale pack sizes.
  • Supplied with product information and a datasheet for laboratory use.