Learn More
Accela Chembio Inc 2-fluoro-3-formyl-4-iodopyridine | 1g | 153034-82-3 | MFCD03095290 | 97+% | Shelf Life: 1980 Days | Light Sensitive/+4/n2
Supplier:  Accela Chembio Inc SY0072241G
Encompass_Preferred
2-fluoro-3-formyl-4-iodopyridine | 1g | 153034-82-3 | MFCD03095290 | 97+% | Shelf Life: 1980 Days | Light Sensitive/+4/n2
Catalog No. 50-251-270