Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-hydroxypyridine, 97%, Thermo Scientific™

Catalog No. AAH32862MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32862MD 250 mg
AAH3286203 1 g
AAH3286206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH32862MD Supplier Thermo Scientific Chemicals Supplier No. H32862MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174669-74-0
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.09
MDL Number MFCD04112569
InChI Key UEQRKEWMEMJXQO-UHFFFAOYSA-N
Synonym 2-fluoro-3-hydroxypyridine, 2-fluoro-3-pyridinol, 2-fluoropyridine-3-ol, 3-pyridinol, 2-fluoro, 2-fluoro-3-hydroxy pyridine, pubchem6290, acmc-209e8o, 3-hydroxy-2-fluoropyridine, 3-pyridinol, 2-fluoro-9ci
PubChem CID 2762785
IUPAC Name 2-fluoropyridin-3-ol
SMILES OC1=C(F)N=CC=C1

Specifications

Melting Point 131°C to 133°C
Quantity 250 mg
Formula Weight 113.09
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-hydroxypyridine
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.