Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Fluoro-3-iodopyridine, 98%, [113975-22-7], F14442 222.99 C5H3FIN

Catalog No. 501409278
Encompass_Preferred

Compound: 2-Fluoro-3-iodopyridine

CAS#: 113975-22-7

MDL#: not listed

Background: A fluorinated, iodinated pyridine used as a building block in pharmaceutical research. The iodine offers a highly reactive site for cross-coupling, while the fluorine supports nucleophilic aromatic substitution chemistry.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-9278 Supplier Frontier Specialty Chemicals Supplier No. F144421G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Fluoro-3-iodopyridine

CAS#: 113975-22-7

MDL#: not listed

Background: A fluorinated, iodinated pyridine used as a building block in pharmaceutical research. The iodine offers a highly reactive site for cross-coupling, while the fluorine supports nucleophilic aromatic substitution chemistry.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.