Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Fluoro-3-methoxyphenylboronic acid | 352303-67-4 | 99.98% | 169.95 | 25 G
SDP

Catalog No. 5000403063
Encompass_Preferred

2-Fluoro-3-methoxyphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular weight: 169.95
  • Formula: C7H8BFO3
  • CAS No.: 352303-67-4
  • Appearance: Solid, white to off-white
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-03063 Supplier Medchemexpress LLC Supplier No. 25GHY32923
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Fluoro-3-methoxyphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular weight: 169.95
  • Formula: C7H8BFO3
  • CAS No.: 352303-67-4
  • Appearance: Solid, white to off-white
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.