Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. F05335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05335G 5 g
F05331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05335G Supplier TCI America Supplier No. F05335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2369-18-8 | C6H6FN | 111.119 g/mol

Chemical Identifiers

CAS 2369-18-8
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041223
InChI Key PHGXUFPYCAWEHK-UHFFFAOYSA-N
Synonym 2-fluoro-3-picoline, 2-fluoro-3-methyl pyridine, 2-fluoro-3-methyl-pyridine, pyridine, 2-fluoro-3-methyl, 2-fluor-3-methylpyridin, pubchem3992, acmc-1ccv4, ksc497i8p, 2-fluoro-3-methylpyridine
PubChem CID 242722
IUPAC Name 2-fluoro-3-methylpyridine
SMILES CC1=C(N=CC=C1)F

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 g
UN Number 1993
Formula Weight 111.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.