Learn More
Indofine Chemical 2-Fluoro-3-(Trifluo, 97%, 5 Gm
SDP
Catalog No. 50913324
Encompass_Preferred
2-FLUORO-3-(TRIFLUOROMETHYL)BENZOYL CHLORIDE 5 GM, Fluoro Organics, C8H3ClF4O, MW: 226.56, 97%, 208173-19-7, MFCD00061154
Catalog No. 50-913-324 Supplier Indofine Chemical Supplier No. 093601 5 GM
2-FLUORO-3-(TRIFLUOROMETHYL)BENZOYL CHLORIDE 5 GM, Fluoro Organics, C8H3ClF4O, MW: 226.56, 97%, 208173-19-7, MFCD00061154