Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F10065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1006-5G 5 g
F10061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1006-5G Supplier TCI America Supplier No. F10065G
Add to Cart
Add to Cart

Specifications

CAS 157834-21-4
Melting Point 58°C
Color White
Molecular Formula C7H5BF4O2
Quantity 5 g
MDL Number MFCD07784336
Synonym 2-fluoro-3-trifluoromethyl phenylboronic acid, 2-fluoro-3-trifluoromethyl benzeneboronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethylphenylboronic acid, 2-fluoro-3-trifluoromethyl phenylboron
InChI Key PKARAEQMGHJHMY-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.919
PubChem CID 16217169
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.