Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-fluoro-4-biphenyl)acetic acid-d5 | 2733149-72-7 | MFCD00143661 | 99.6% | 235.27 g·mol⁻¹ | C14H6D5FO2 | 5 MG
SDP

Catalog No. 5000221466
Encompass_Preferred

(2-Fluoro-4-biphenyl)acetic acid-d5 is the deuterium-labeled analogue of (2-Fluoro-4-biphenyl)acetic acid used as a stable isotope internal standard or tracer for quantitative analytical methods. Supplied as a powder with defined purity and specified storage recommendations for long-term stability.

  • Deuterium-labeled analogue for use as an internal standard in quantitative NMR and mass spectrometry.
  • High purity (≈99.6%) suitable for analytical reference.
  • Molecular weight 235.27 g·mol⁻¹ and formula C14H6D5FO2.
  • Available in small vials (e.g., 5 mg) for trace-level analyses.
  • Powder form with recommended storage at -20°C for multi-year stability.

Catalog No. 50-002-21466 Supplier Medchemexpress LLC Supplier No. HY142907S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Fluoro-4-biphenyl)acetic acid-d5 is the deuterium-labeled analogue of (2-Fluoro-4-biphenyl)acetic acid used as a stable isotope internal standard or tracer for quantitative analytical methods. Supplied as a powder with defined purity and specified storage recommendations for long-term stability.

  • Deuterium-labeled analogue for use as an internal standard in quantitative NMR and mass spectrometry.
  • High purity (≈99.6%) suitable for analytical reference.
  • Molecular weight 235.27 g·mol⁻¹ and formula C14H6D5FO2.
  • Available in small vials (e.g., 5 mg) for trace-level analyses.
  • Powder form with recommended storage at -20°C for multi-year stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.