Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific™

Catalog No. AC263540050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263540050 5 g Glass bottle
AC263540250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC263540050 Supplier Thermo Scientific Chemicals Supplier No. 263540050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 74457-86-6
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00042290
InChI Key PIRRWUMTIBFCCW-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
PubChem CID 592821
IUPAC Name 1-(2-fluoro-4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C(C)=O)C(F)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 54.0°C
Color Colorless
Boiling Point 80.0°C to 85.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 168.17
Percent Purity 99%
Physical Form Viscous Solution
Chemical Name or Material 2'-Fluoro-4'-methoxyacetophenone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.