Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 2-Fluoro-4-methoxybenzaldehyde, 331-64-6, MFCD00236679, 5g

Catalog No. 501473352
Encompass_Preferred

Molecular Formula C8H7FO2, Purity 98%, Molecular Weight 154.14, Melting Point 47°

Catalog No. 50-147-3352 Supplier Matrix Scientific Supplier No. 0025545G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C8H7FO2, Purity 98%, Molecular Weight 154.14, Melting Point 47°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.