Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Fluoro-4-methyl-5-(tributylstannyl)pyridine

Catalog No. SYX000851G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX000851G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX000851G Supplier Sigma Aldrich Supplier No. SYX000851G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methylcyclohexene is used in the synthesis of an enantiomerically pure oxaliplatin derivative, {(1R,2R)-4-methyl-1,2-cyclohexanediamine}oxalatoplatinum(ii), which exhibits good anticancer activity
  • It is also used in the preparation of n,n-diethyldithiocarbamate functionalized 1,4-polyisoprenes

Specifications

CAS 1245816-06-1
Molecular Formula C18H32FNSn
Linear Formula C18H32FNSn
MDL Number MFCD12827558
Quantity 5 g
Molecular Weight (g/mol) 400.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.