Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH62709MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62709MD 250 mg
AAH6270903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62709MD Supplier Thermo Scientific Chemicals Supplier No. H62709MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-68-0
Molecular Formula C13H15BF4O2
Molecular Weight (g/mol) 290.06
MDL Number MFCD08458187
InChI Key ZXEYGQNXUCXTBK-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethylphenylboronic acid, pinacol ester, 2-2-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl-phenylboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl benzeneboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl phenyl boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-2-fluoro-4-trifluoromethyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 17750248
IUPAC Name 2-[2-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.