Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™

Catalog No. AAH2657306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2657306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2657306 Supplier Thermo Scientific Chemicals Supplier No. H2657306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in the field of nanoparticles. Used in laboratory as reagent.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209991-64-0
Molecular Formula C9H6F4O2
Molecular Weight (g/mol) 222.14
MDL Number MFCD00061186
InChI Key LUIOBGWYVHPKRC-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl phenylacetic acid, 2-fluoro-4-trifluoromethyphenylacetic acid, 2-2-fluoro-4-trifluoromethyl phenyl acetic acid, 2-fluoro-4-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 2-fluoro-4-trifluoromethyl, pubchem2710, pubchem8433, 2-fluoro-4-trifluoromethyphenylaceticacid, acmc-1cisp
PubChem CID 3863567
IUPAC Name 2-[2-fluoro-4-(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=C(F)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 78°C to 81°C
Quantity 5 g
Formula Weight 222.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.