Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH62689MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62689MD 250 mg
AAH6268903 1 g
AAH6268906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62689MD Supplier Thermo Scientific Chemicals Supplier No. H62689MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112208-82-8
Molecular Formula C13H16BFO3
Molecular Weight (g/mol) 250.08
MDL Number MFCD11867877
InChI Key WHYMLQVABFAGEK-UHFFFAOYSA-N
Synonym 2-fluoro-5-formylphenylboronic acid pinacol ester, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-fluoro-5-formylphenylboronic acid, pinacol ester, 2-fluoro-5-formylphenyl boronic acid pinacol ester, 2-fluoro-5-formylbenzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzaldehyde, 4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzaldehyde, benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 46738018
IUPAC Name 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=CC(C=O)=C1

Specifications

Melting Point 35°C to 40°C
Flash Point >110°C (230°F)
Quantity 250 mg
Formula Weight 250.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-5-formylbenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.