Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Fluoro-5-formylbenzonitrile | 218301-22-5 | 99.7% | 149.12 | 50 G
SDP

Catalog No. 5000404793
Encompass_Preferred

2-Fluoro-5-formylbenzonitrile is a drug intermediate for the synthesis of various active compounds.

  • Suitable for synthesis of various active compounds.
  • Appearance: solid.
  • Color: off-white to light yellow.
  • Shipping: room temperature in continental US.
  • Storage: 4°C, stored under nitrogen.

Catalog No. 50-004-04793 Supplier Medchemexpress LLC Supplier No. HY2013850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Fluoro-5-formylbenzonitrile is a drug intermediate for the synthesis of various active compounds.

  • Suitable for synthesis of various active compounds.
  • Appearance: solid.
  • Color: off-white to light yellow.
  • Shipping: room temperature in continental US.
  • Storage: 4°C, stored under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.