Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F09151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09151G 1 g
F0915-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09151G Supplier TCI America Supplier No. F09151G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Fluoro-5-methylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 166328-16-1
Color White-Yellow
Molecular Formula C7H8BFO2
Quantity 1 g
MDL Number MFCD03428512
Synonym 2-fluoro-5-methylbenzeneboronic acid, 2-fluoro-5-methylphenyl boronic acid, 2-fluoro-5-methylphenyl boranediol, 2-fluoro-5-methylphenylboronic, boronic acid, 2-fluoro-5-methylphenyl, 6-fluoro-m-tolylboronic acid, boronic acid, 2-fluoro-5-methylphenyl-9ci, 2-fluoro-5-methyl-phenyl boronic acid, pubchem23774, acmc-1bybh
InChI Key ZLODKAZZRDLUKX-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)C)F)(O)O
IUPAC Name (2-fluoro-5-methylphenyl)boronic acid
Molecular Weight (g/mol) 153.947
PubChem CID 10351884
Formula Weight 153.95
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.