Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. F09835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09835G 5 g
F098325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09835G Supplier TCI America Supplier No. F09835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 456-24-6 | C5H3FN2O2 | 142.089 g/mol

Chemical Identifiers

CAS 456-24-6
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.089
MDL Number MFCD03095059
InChI Key XOZAJNLUAODXSP-UHFFFAOYSA-N
Synonym 6-fluoro-3-nitropyridine, pyridine, 2-fluoro-5-nitro, 2-fluor-5-nitropyridin, 2-fluoro-5-nitro-pyridine, abbypharma ap-15-5253, pubchem1202, acmc-1arnr, 5-nitro-2-fluoropyridine, 2-fluoro-5-nitro pyridine
PubChem CID 95264
IUPAC Name 2-fluoro-5-nitropyridine
SMILES C1=CC(=NC=C1[N+](=O)[O-])F

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 87°C
Quantity 5 g
Formula Weight 142.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.