Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™
SDP

Catalog No. F02891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02891G 1 g
F02895G 5 g
F028925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F02891G Supplier TCI America Supplier No. F02891G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146137-78-2
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.11
MDL Number MFCD00061311
InChI Key IDLNLGMUINCSGS-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl benzaldehyde, 2-fluoro-5-trifluoromethyl-benzaldehyde, 4-fluoro-3-formylbenzotrifluoride, benzaldehyde, 2-fluoro-5-trifluoromethyl, pubchem5798, acmc-209cwf, intermediates-zcf02118, ksc496a6r
PubChem CID 2734904
IUPAC Name 2-fluoro-5-(trifluoromethyl)benzaldehyde
SMILES FC1=CC=C(C=C1C=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 192.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.