Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-(trifluoromethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. F08605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08605G 5 g
F08601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08605G Supplier TCI America Supplier No. F08605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4088-84-0
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00061280
InChI Key LCLVMSCLLULGRY-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl benzonitrile, 3-cyano-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-tetrafluoro-m-tolunitrile, benzonitrile, 2-fluoro-5-trifluoromethyl, 2-fluoro-5-trifluoromethyl benzenecarbonitrile, pubchem2739, ksc583s5h, 3-cyano-4-fluorotrifluorotoluene, 2-fluoro-5-trifluoromethyl-benzonitrile
PubChem CID 521016
IUPAC Name 2-fluoro-5-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)C#N)F

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
UN Number 3276
Formula Weight 189.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.