Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F07885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0788-5G 5 g
F07881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0788-5G Supplier TCI America Supplier No. F07885G
Add to Cart
Add to Cart

Specifications

CAS 352535-96-7
Melting Point 109°C
Color White
Molecular Formula C7H5BF4O2
Quantity 5 g
MDL Number MFCD03701538
Synonym 2-fluoro-5-trifluoromethyl phenylboronic acid, 2-fluoro-5-trifluoromethyl phenyl boronic acid, 2-fluoro-5-trifluoromethyl benzeneboronic acid, 2-fluoro-5-trifluoromethyl boronic acid, 2-fluoro-5-trifluoromethylphenylboronic acid
InChI Key KUHVVLFCTMTYGR-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-5-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.919
PubChem CID 2763301
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.