Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. F08465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08465G 5 g
F08461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08465G Supplier TCI America Supplier No. F08465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 69045-82-5 | C6H3F4N | 165.091 g/mol

Chemical Identifiers

CAS 69045-82-5
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD07437945
InChI Key UUODQIKUTGWMPT-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-5-trifluoromethyl-pyridine, pyridine, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-3-picoline, pubchem9252, acmc-209o5o, ksc628s4n
PubChem CID 5463754
IUPAC Name 2-fluoro-5-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
UN Number 1993
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.