Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-fluoro-6-[[(3S)-oxolan-3-yl]amino]-4-(3,6,6-trimethyl-4-oxo-5,7-dihydroindol-1-yl)benzamide | 1073969-18-2 | 99.1% | 399.46 | C22H26FN3O3 | 100 MG
SDP

Catalog No. 5000307613
Encompass_Preferred

SNX-0723 is a small-molecule Hsp90 inhibitor (CAS 1073969-18-2) supplied for research use. It exhibits high binding affinity for human and Plasmodium falciparum Hsp90 and shows activity in cellular and parasite assays.

  • Purity 99.1% by HPLC.
  • Molecular formula C22H26FN3O3, molecular weight 399.46 Da.
  • Soluble in DMSO (20.83 mg/mL) and suitable for formulation for in vivo studies.
  • HsHsp90 Ki = 4.4 nM; PfHsp90 Ki = 47 nM; P. berghei EC50 = 3.3 μM.
  • Available as a 100 mg solid pack for research applications.

Catalog No. 50-003-07613 Supplier Medchemexpress LLC Supplier No. HY119046100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SNX-0723 is a small-molecule Hsp90 inhibitor (CAS 1073969-18-2) supplied for research use. It exhibits high binding affinity for human and Plasmodium falciparum Hsp90 and shows activity in cellular and parasite assays.

  • Purity 99.1% by HPLC.
  • Molecular formula C22H26FN3O3, molecular weight 399.46 Da.
  • Soluble in DMSO (20.83 mg/mL) and suitable for formulation for in vivo studies.
  • HsHsp90 Ki = 4.4 nM; PfHsp90 Ki = 47 nM; P. berghei EC50 = 3.3 μM.
  • Available as a 100 mg solid pack for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.