Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-(trifluoromethyl)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. F10335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10335G 5 g
F10331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10335G Supplier TCI America Supplier No. F10335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60611-24-7
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.11
MDL Number MFCD00061273
InChI Key FAKUGVHRTLCKHB-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-6-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-o-tolualdehyde, pubchem4236, 2-fluoro-6-trifluoromethyl-benzaldehyde, acmc-1b5by, otf-bad-6f, 3-fluoro-2-formylbenzotrifluoride, attercop-chm at111495
PubChem CID 521933
IUPAC Name 2-fluoro-6-(trifluoromethyl)benzaldehyde
SMILES FC1=CC=CC(=C1C=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 g
Formula Weight 192.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.