Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. F07571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07571G 1 g
F07575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07571G Supplier TCI America Supplier No. F07571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32890-94-1
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00040981
InChI Key LNARMXLVVGHCRP-UHFFFAOYSA-M
Synonym 2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626
PubChem CID 688123
IUPAC Name 2-fluoro-6-(trifluoromethyl)benzoate
SMILES [O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F

Specifications

Melting Point 88°C
Color White
Quantity 1 g
Formula Weight 208.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.