Learn More
eMolecules​ 136834-22-5 | DMT-2'fluoro-da(bz) amidite | ChemScene | MFCD12912392 | 875.939 | C47H51FN7O7P | 95.000 | COc1ccc(cc1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1OP(OCCC#N)N(C(C)C)C(C)C)n1cnc2c(NC(=O)c3ccccc3)ncnc12)(c1ccccc1)c1ccc(OC)cc1 | 100g | 941423346
Supplier:  eMolecules​ 941423346
Encompass_Preferred
DMT-2'fluoro-da(bz) amidite | ChemScene | 136834-22-5 | MFCD12912392 | 875.939 | C47H51FN7O7P | 95.000 | COc1ccc(cc1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1OP(OCCC#N)N(C(C)C)C(C)C)n1cnc2c(NC(=O)c3ccccc3)ncnc12)(c1ccccc1)c1ccc(OC)cc1 | 100g | 941423346
Catalog No. NC3971605