Learn More
Cayman Chemical 2-fluoro Palmitic Acid, Purity: ≥95%, CAS: 16518-94-8, 10 mg
SDP
Catalog No. 502892873
Encompass_Preferred
2-fluoro Palmitic acid inhibits sphingosine biosynthesis and long-chain acyl-CoA synthetase with an IC50 value of 0.2 mM.{1613}
Catalog No. 50-289-2873 Supplier Cayman Chemical Supplier No. 9038010 MG
2-fluoro Palmitic acid inhibits sphingosine biosynthesis and long-chain acyl-CoA synthetase with an IC50 value of 0.2 mM.{1613}