Learn More
Cayman Chemical 2-fluoro PalmItIc AcId 25mg
SDP
Catalog No. 502895004
Encompass_Preferred
2-fluoro Palmitic acid inhibits sphingosine biosynthesis and long-chain acyl-CoA synthetase with an IC50 value of 0.2 mM.{1613}
Catalog No. 50-289-5004 Supplier Cayman Chemical Supplier No. 9038025 MG
2-fluoro Palmitic acid inhibits sphingosine biosynthesis and long-chain acyl-CoA synthetase with an IC50 value of 0.2 mM.{1613}