Learn More
Cayman Chemical 2-Fluoroadenine, Purity: ≥98%, CAS: 700-49-2, 500 mg
SDP
Catalog No. 502991212
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of Hsp90 inhibitors that have anticancer activity in vitro; cytotoxic to CEM human leukemia cells (IC50 = 0.15 µM)
Catalog No. 50-299-1212 Supplier Cayman Chemical Supplier No. 30421500 MG
A heterocyclic building block; has been used in the synthesis of Hsp90 inhibitors that have anticancer activity in vitro; cytotoxic to CEM human leukemia cells (IC50 = 0.15 µM)