Learn More
Medchemexpress LLC 2-Fluoroadenine | 700-49-2 | 153.12 | 250 MG
SDP
Catalog No. 5000449958
Encompass_Preferred
2-Fluoroadenine is a toxic purine base that exhibits toxicity in both nonproliferating and proliferating tumor cells. It can be used for anticancer research.
  • Toxic purine base
  • Targets DNA/RNA synthesis
  • Part of the Cell Cycle/DNA Damage pathway
  • Inhibits cell growth by targeting enzymes involved in RNA or protein synthesis
  • Exhibits cytotoxicity in nonproliferating tumor cells
  • Inhibits protein, RNA, and DNA syntheses in Balb-3T3 cells
  • Used for anticancer research
Catalog No. 50-004-49958 Supplier Medchemexpress LLC Supplier No. 250MGHYW008469
2-Fluoroadenine is a toxic purine base that exhibits toxicity in both nonproliferating and proliferating tumor cells. It can be used for anticancer research.
  • Toxic purine base
  • Targets DNA/RNA synthesis
  • Part of the Cell Cycle/DNA Damage pathway
  • Inhibits cell growth by targeting enzymes involved in RNA or protein synthesis
  • Exhibits cytotoxicity in nonproliferating tumor cells
  • Inhibits protein, RNA, and DNA syntheses in Balb-3T3 cells
  • Used for anticancer research