Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzamide 98%, Thermo Scientific™

Catalog No. AC153290250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC153290250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC153290250 Supplier Thermo Scientific Chemicals Supplier No. 153290250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 445-28-3
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6FNO
Linear Formula FC6H4CONH2
Quantity 25g
MDL Number MFCD00007970
Synonym o-fluorobenzamide, benzamide, 2-fluoro, benzamide, o-fluoro, o-fluorobenzoic acid amide, ortho-fluorobenzamide, fluorobenzamide, 2-fluoro-benzamide, pubchem3405, acmc-1aljb, 2-fluorobenzamide
InChI Key KGGHWIKBOIQEAJ-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(=O)N)F
IUPAC Name 2-fluorobenzamide
Molecular Weight (g/mol) 139.129
PubChem CID 67964
Formula Weight 139.13
Percent Purity ≥97.5%
Chemical Name or Material 2-Fluorobenzamide, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-157-6
RTECSNumber CV4953333
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.