Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzamide 98.0+%, TCI America™
SDP

Catalog No. F027810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F027810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F027810G Supplier TCI America Supplier No. F027810G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 445-28-3
Molecular Formula C7H6FNO
Molecular Weight (g/mol) 139.129
MDL Number MFCD00007970
InChI Key KGGHWIKBOIQEAJ-UHFFFAOYSA-N
Synonym o-fluorobenzamide, benzamide, 2-fluoro, benzamide, o-fluoro, o-fluorobenzoic acid amide, ortho-fluorobenzamide, fluorobenzamide, 2-fluoro-benzamide, pubchem3405, acmc-1aljb, 2-fluorobenzamide
PubChem CID 67964
IUPAC Name 2-fluorobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)F

Specifications

Melting Point 117°C
Color White
Quantity 10 g
Formula Weight 139.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorobenzamide
RTECSNumber CV4953333
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.