Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 103°C |
| Quantity | 25 g |
| Formula Weight | 121.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Fluorobenzonitrile |
Sécurité et manipulation
| TSCA | Yes |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'
