Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzotrichloride 98.0+%, TCI America™
SDP

Catalog No. F02905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02905G Supplier TCI America Supplier No. F02905G
Add to Cart
Add to Cart

Specifications

CAS 488-98-2
Color Yellow
Boiling Point 75°C
Molecular Formula C7H4Cl3F
Quantity 5 g
MDL Number MFCD00000787
Synonym 2-Fluoro-alpha,alpha,alpha-trichlorotoluene
InChI Key JYLVSNNSOWDQQX-UHFFFAOYSA-N
SMILES FC1=CC=CC=C1C(Cl)(Cl)Cl
IUPAC Name 1-fluoro-2-(trichloromethyl)benzene
Molecular Weight (g/mol) 213.46
PubChem CID 68104
Formula Weight 213.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzotrichloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.