Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzyl bromide, 98%

Catalog No. AC197960050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197960050 5 g Glass bottle
AC197960250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197960050 Supplier Thermo Scientific Chemicals Supplier No. 197960050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 446-48-0 | C7H6BrF | 189.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-48-0
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.03
MDL Number MFCD00000324
InChI Key FFWQLZFIMNTUCZ-UHFFFAOYSA-N
Synonym 2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide
PubChem CID 67968
IUPAC Name 1-(bromomethyl)-2-fluorobenzene
SMILES C1=CC=C(C(=C1)CBr)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5670g/mL
Boiling Point 84°C to 85°C (15.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5495 to 1.5515
Linear Formula FC6H4CH2Br
Quantity 5 g
Beilstein 05, II, 238
Specific Gravity 1.567
Formula Weight 189.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzyl bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-169-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.