Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorocyclohexanol 98.0+%, TCI America™
SDP

Catalog No. F06315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06315G 5 g
F06311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06315G Supplier TCI America Supplier No. F06315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 656-60-0
Molecular Formula C6H11FO
Molecular Weight (g/mol) 118.151
MDL Number MFCD08146648
InChI Key LMYKFDDTPIOYQV-PHDIDXHHSA-N
Synonym trans-2-fluorocyclohexanol, 1r,2r-2-fluorocyclohexan-1-ol, r,r---2-fluorocyclohexanol, trans-2-fluoro-cyclohexanol, 2alpha-fluorocyclohexan-1beta-ol, cyclohexanol, 2-fluoro-, 1r,2r, trans-2-fluorocyclohexanol unspecified stereochemistry
PubChem CID 12700773
IUPAC Name (1R,2R)-2-fluorocyclohexan-1-ol
SMILES C1CCC(C(C1)O)F

Specifications

Melting Point 21°C
Color White-Yellow
Boiling Point 166°C
Quantity 5 g
Formula Weight 118.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluorocyclohexanol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.