Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenethyl alcohol, 99%, Thermo Scientific™

Catalog No. AC310970250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310970250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC310970250 Supplier Thermo Scientific Chemicals Supplier No. 310970250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50919-06-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00002887
InChI Key HNIGZVZDWCTFPR-UHFFFAOYSA-N
Synonym 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol
PubChem CID 2733296
IUPAC Name 2-(2-fluorophenyl)ethanol
SMILES OCCC1=CC=CC=C1F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0400g/mL
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4CH2CH2OH
Refractive Index 1.5080 to 1.51
Quantity 25 g
Specific Gravity 1.04
Formula Weight 140.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenethyl alcohol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.