Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Fluorophenol

Catalog No. F12804100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F12804100G 100 g
F1280410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F12804100G Supplier Sigma Aldrich Supplier No. F12804100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Fluorophenol

Specifications

CAS 367-12-4
Melting Point 16.1°C (lit.)
Density 1.256 g/mL (at 25°C (literature))
Boiling Point 171°C to 172°C (741 mmHg)
Molecular Formula C6H5FO
Refractive Index n20/D 1.511 (literature)
Linear Formula FC6H4OH
MDL Number MFCD00002155
Quantity 100 g
Molecular Weight (g/mol) 112.1
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.