Learn More
Medchemexpress LLC 2-Fluorophenol | 367-12-4 | 99.96% | 112.10 | 250 G
SDP
Catalog No. 5000402518
Encompass_Preferred
2-Fluorophenol is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Appears colorless to light yellow
  • Exists as a solid below 16.1°C and a liquid above 16.1°C
Catalog No. 50-004-02518 Supplier Medchemexpress LLC Supplier No. 250GHY41755
2-Fluorophenol is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Appears colorless to light yellow
  • Exists as a solid below 16.1°C and a liquid above 16.1°C