Learn More
Medchemexpress LLC 2-Fluorophenylboronic acid | 1993-03-9 | 139.92 | 25 G
SDP
Catalog No. 5000449964
Encompass_Preferred
2-Fluorophenylboronic acid is a versatile compound intermediate, characterized as a white to off-white solid with a molecular weight of 139.92. It is highly soluble in DMSO at 100 mg/mL, though hygroscopic DMSO requires careful handling.
  • Used to functionalize IPG
  • Employed in the synthesis of compounds with ABS activity
  • Utilized in P. falciparum infection studies
  • Molecular formula: C6H6BFO2
Powder can be stored at -20°C for 3 years or 4°C for 2 years. In solvent, store at -80°C for 6 months or -20°C for 1 month. Aliquoted solutions should be stored to prevent inactivation from repeated freeze-thaw cycles.
Catalog No. 50-004-49964 Supplier Medchemexpress LLC Supplier No. 25GHYW001240
Add to Cart
Add to Cart
2-Fluorophenylboronic acid is a versatile compound intermediate, characterized as a white to off-white solid with a molecular weight of 139.92. It is highly soluble in DMSO at 100 mg/mL, though hygroscopic DMSO requires careful handling.
  • Used to functionalize IPG
  • Employed in the synthesis of compounds with ABS activity
  • Utilized in P. falciparum infection studies
  • Molecular formula: C6H6BFO2
Powder can be stored at -20°C for 3 years or 4°C for 2 years. In solvent, store at -80°C for 6 months or -20°C for 1 month. Aliquoted solutions should be stored to prevent inactivation from repeated freeze-thaw cycles.