Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F04075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0407-5G 5 g
F0407-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0407-5G Supplier TCI America Supplier No. F04075G
Add to Cart
Add to Cart

Specifications

CAS 1993-03-9
Melting Point 110°C
Molecular Formula C6H6BFO2
Quantity 5 g
MDL Number MFCD00674013
Synonym 2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl
InChI Key QCSLIRFWJPOENV-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1F)(O)O
IUPAC Name (2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 139.92
PubChem CID 2734354
Formula Weight 139.92
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.