Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine-4-boronic acid, 98%

Catalog No. AC448730050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448730050 5 g Glass bottle
AC448730010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448730050 Supplier Thermo Scientific Chemicals Supplier No. 448730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 401815-98-3 | C5H5BFNO2 | 140.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401815-98-3
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
MDL Number MFCD04112534
InChI Key WXGBZJJAGLSBPR-UHFFFAOYSA-N
Synonym 2-fluoropyridine-4-boronic acid, 2-fluoropyridin-4-yl boronic acid, 2-fluoro-4-pyridineboronic acid, 2-fluoro-4-pyridinylboronic acid, 2-fluoropyridine-4-boronicacid, 2-fluoropyridin-4-yl-4-boronic acid, 2-fluoropyridine-boronic acid, 2-fluoro-4-pyridyl boronic acid, 2-fluoro-pyridine-4-boronic acid, boronic acid, 2-fluoro-4-pyridinyl
PubChem CID 2783396
IUPAC Name (2-fluoropyridin-4-yl)boronic acid
SMILES OB(O)C1=CC=NC(F)=C1

Specifications

Melting Point 212°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 140.91
Percent Purity 98%
Chemical Name or Material 2-Fluoropyridine-4-boronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.