Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193963 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09351G 1 g
F0935-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09351G Supplier TCI America Supplier No. F09351G
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Fluoropyridine-4-boronic Acid (contains varying amounts of Anhydride)
CAS 401815-98-3
Melting Point 200°C
Color White
Molecular Formula C5H5BFNO2
MDL Number MFCD04112534
Quantity 1 g
Synonym 2-fluoropyridine-4-boronic acid, 2-fluoropyridin-4-yl boronic acid, 2-fluoro-4-pyridineboronic acid, 2-fluoro-4-pyridinylboronic acid, 2-fluoropyridine-4-boronicacid, 2-fluoropyridin-4-yl-4-boronic acid, 2-fluoropyridine-boronic acid, 2-fluoro-4-pyridyl boronic acid, 2-fluoro-pyridine-4-boronic acid, boronic acid, 2-fluoro-4-pyridinyl
InChI Key WXGBZJJAGLSBPR-UHFFFAOYSA-N
SMILES OB(O)C1=CC=NC(F)=C1
IUPAC Name (2-fluoropyridin-4-yl)boronic acid
Molecular Weight (g/mol) 140.91
PubChem CID 2783396
Formula Weight 140.91
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.