Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine-4-boronic acid pinacol ester, 95%

Catalog No. AC435930050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435930050 5 g Glass bottle
AC435930010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435930050 Supplier Thermo Scientific Chemicals Supplier No. 435930050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 458532-86-0 | C11H15BFNO2 | 223.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-86-0
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.05
MDL Number MFCD06798253
InChI Key PCLMNCBIXQQRMB-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12060164
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 47.9°C to 51.6°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 223.05
Percent Purity 95%
Physical Form Crystals or Powder
Chemical Name or Material 2-Fluoropyridine-4-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.