Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 2-(Fmoc-amino)-4-cyclopropylbutanoic acid | 10 g | CAS 1694007-54-9 | InChIKey RFARSMCKSWZMRL-UHFFFAOYSA-N

Catalog No. 502357725
Encompass

2-(Fmoc-amino)-4-cyclopropylbutanoic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1694007-54-9
- MDL: No data
- InChIKey: RFARSMCKSWZMRL-UHFFFAOYSA-N
- Molecular Weight: 365.429
- Molecular Formula: C22H23NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-cyclopropylbutanoic acid
- SMILES: O=C(C(CCC1CC1)NC(OCC2C3=CC=CC=C3C4=CC=CC=C24)=O)O

Catalog No. 50-235-7725 Supplier STA Pharmaceutical Supplier No. STAAA1097210GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Fmoc-amino)-4-cyclopropylbutanoic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1694007-54-9
- MDL: No data
- InChIKey: RFARSMCKSWZMRL-UHFFFAOYSA-N
- Molecular Weight: 365.429
- Molecular Formula: C22H23NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-cyclopropylbutanoic acid
- SMILES: O=C(C(CCC1CC1)NC(OCC2C3=CC=CC=C3C4=CC=CC=C24)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.